missing translation for 'onlineSavingsMsg'
Learn More

2-(Trimethylsilylethoxymethyl)triphenylphosphonium chloride, 97%, Thermo Scientific™

Catalog Number 11945881
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
82495-75-8
C24H30ClOPSi
429.01
NEUMNYXEDIPGJD-UHFFFAOYSA-M
2-trimethylsilyl ethoxymethyltriphenylphosphonium chloride, sem-triphenylphosphonium chloride, 2-trimethylsilyl ethoxymethyl triphenylphosphonium chloride, phosphonium, triphenyl 2-trimethylsilyl ethoxy methyl-, chloride, acmc-209pp9, 2-trimethylsilyl ethoxymethyltriphenylphosphoniumchloride, 2-trimethylsilyl ethoxymethyl-triphenylphosphonium chloride, 2-trimethylsilyl-ethoxymethyl triphenylphosphonium chloride, triphenyl-2-trimethylsilanyl-ethoxymethyl phosphonium chloride
11396265
triphenyl(2-trimethylsilylethoxymethyl)phosphanium;chloride
C[Si](C)(C)CCOC[P+](C1=CC=CC=C1)(C2=CC=CC=C2)C3=CC=CC=C3.[Cl-]
This item is not returnable. View return policy
82495-75-8
C24H30ClOPSi
429.01
NEUMNYXEDIPGJD-UHFFFAOYSA-M
2-trimethylsilyl ethoxymethyltriphenylphosphonium chloride, sem-triphenylphosphonium chloride, 2-trimethylsilyl ethoxymethyl triphenylphosphonium chloride, phosphonium, triphenyl 2-trimethylsilyl ethoxy methyl-, chloride, acmc-209pp9, 2-trimethylsilyl ethoxymethyltriphenylphosphoniumchloride, 2-trimethylsilyl ethoxymethyl-triphenylphosphonium chloride, 2-trimethylsilyl-ethoxymethyl triphenylphosphonium chloride, triphenyl-2-trimethylsilanyl-ethoxymethyl phosphonium chloride
11396265
triphenyl(2-trimethylsilylethoxymethyl)phosphanium;chloride
C[Si](C)(C)CCOC[P+](C1=CC=CC=C1)(C2=CC=CC=C2)C3=CC=CC=C3.[Cl-]
Melting Point 166.0°C
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula (CH3)3Si(CH2)3OCH2P(C6H5)3Cl
Quantity 25 g
Formula Weight 429.01
Percent Purity 97%
Chemical Name or Material 2-(Trimethylsilylethoxymethyl)triphenylphosphonium chloride
compliance-icons
Product Identifier
  • 2-(Trimethylsilylethoxymethyl)triphenylphosphonium chloride
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P402+P404-Store in a dry place. Store in a closed container.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'