missing translation for 'onlineSavingsMsg'
Learn More

2-[(Trimethylsilyl)ethynyl]aniline, 97%, Thermo Scientific Chemicals

Catalog Number 15472328
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
103529-16-4
C11H15NSi
189.333
MFCD02093646
ISXSBRZDPKQYSC-UHFFFAOYSA-N
2-trimethylsilyl ethynyl aniline, 2-2-trimethylsilylethynyl aniline, 2-trimethylsilylethynyl aniline, 2-2-trimethylsilyl ethynyl aniline, benzenamine, 2-trimethylsilyl ethynyl
4377953
2-(2-trimethylsilylethynyl)aniline
C[Si](C)(C)C#CC1=CC=CC=C1N
This item is not returnable. View return policy
103529-16-4
C11H15NSi
189.333
MFCD02093646
ISXSBRZDPKQYSC-UHFFFAOYSA-N
2-trimethylsilyl ethynyl aniline, 2-2-trimethylsilylethynyl aniline, 2-trimethylsilylethynyl aniline, 2-2-trimethylsilyl ethynyl aniline, benzenamine, 2-trimethylsilyl ethynyl
4377953
2-(2-trimethylsilylethynyl)aniline
C[Si](C)(C)C#CC1=CC=CC=C1N

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Colorless to Yellow
Density 0.94 g/mL
Boiling Point 120°C (0.1 mmHg)
Flash Point 110°C (230°F)
Refractive Index 1.564
Quantity 1 g
Formula Weight 189.33
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2-[(Trimethylsilyl)ethynyl]aniline

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'