missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(Trimethylsilyl)ethanol, 98+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
Description
2-(Trimethylsilyl)ethanol is used as a protecting reagent for carboxyl, phosphoryl, hydroxyl and amino group in organic synthesis. It is used as a precursor to prepare trimethyl(2-phenoxyethyl)silanes by reacting with aromatic fluoride. It is also used in the synthesis of teoc-protected amines by using the corresponding isocyanates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-(Trimethylsilyl)ethanol is used as a protecting reagent for carboxyl, phosphoryl, hydroxyl and amino group in organic synthesis. It is used as a precursor to prepare trimethyl(2-phenoxyethyl)silanes by reacting with aromatic fluoride. It is also used in the synthesis of teoc-protected amines by using the corresponding isocyanates.
Solubility
Miscible with water.
Notes
Keep away from sources of ignition. Keep container tightly closed in a dry and well-ventilated place.
2-(Trimethylsilyl)ethanol is used as a protecting reagent for carboxyl, phosphoryl, hydroxyl and amino group in organic synthesis. It is used as a precursor to prepare trimethyl(2-phenoxyethyl)silanes by reacting with aromatic fluoride. It is also used in the synthesis of teoc-protected amines by using the corresponding isocyanates.
Solubility
Miscible with water.
Notes
Keep away from sources of ignition. Keep container tightly closed in a dry and well-ventilated place.
Specifications
Density | 0.825 |
Boiling Point | 71°C to 72°C (10 mm) |
Flash Point | 50°C (122°F) |
Assay Percent Range | 98+% |
Refractive Index | 1.425 |
Quantity | 5 g |
UN Number | UN1987 |
Beilstein | 1732034 |
Solubility Information | Miscible with water. |
Formula Weight | 118.25 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'