missing translation for 'onlineSavingsMsg'
Learn More

2-(Trifluoromethyl)thioxanthen-9-one, 98%, Thermo Scientific™

Catalog Number p-7056088
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
1693-28-3
C14H7F3OS
280.26
MFCD00079785
NEWRXGDGZGIHIS-UHFFFAOYSA-N
2-trifluoromethyl thioxanthen-9-one, 2-trifluoromethyl thioxanthone, 2-trifluoromethyl-9h-thioxanthen-9-one, 2-trifluoromethylthioxanthone, 9h-thioxanthen-9-one, 2-trifluoromethyl, 2-trifludromethylthioxanthone, 2-trifluormethyl-9-thiaxanthen-9-one, 2-trifluoromethyl dibenzo b,e thiin-10-one, pubchem10678, acmc-209dzj
74324
FC(F)(F)C1=CC=C2SC3=CC=CC=C3C(=O)C2=C1
This item is not returnable. View return policy
1693-28-3
C14H7F3OS
280.26
MFCD00079785
NEWRXGDGZGIHIS-UHFFFAOYSA-N
2-trifluoromethyl thioxanthen-9-one, 2-trifluoromethyl thioxanthone, 2-trifluoromethyl-9h-thioxanthen-9-one, 2-trifluoromethylthioxanthone, 9h-thioxanthen-9-one, 2-trifluoromethyl, 2-trifludromethylthioxanthone, 2-trifluormethyl-9-thiaxanthen-9-one, 2-trifluoromethyl dibenzo b,e thiin-10-one, pubchem10678, acmc-209dzj
74324
FC(F)(F)C1=CC=C2SC3=CC=CC=C3C(=O)C2=C1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 147°C to 151°C
Color Yellow
Quantity 1 g
Beilstein 1583738
Sensitivity Light sensitive
IUPAC Name 2-(trifluoromethyl)-9H-thioxanthen-9-one
Formula Weight 280.26
Percent Purity 98%
Physical Form Crystalline
Chemical Name or Material 2-(Trifluoromethyl)thioxanthen-9-one
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'