missing translation for 'onlineSavingsMsg'
Learn More

2-(Trifluoromethyl)quinoline, 97%, Thermo Scientific™

Catalog Number p-4228934
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
347-42-2
C10H6F3N
197.16
MFCD01862002
YZSRICFIQLVSMQ-UHFFFAOYSA-N
2-trifluoromethyl quinoline, 2-trifluoromethyl-quinoline, pubchem23495, trifluoromethyl quinoline, quinoline,2-trifluoromethyl
2777805
2-(trifluoromethyl)quinoline
FC(F)(F)C1=NC2=CC=CC=C2C=C1
This item is not returnable. View return policy
347-42-2
C10H6F3N
197.16
MFCD01862002
YZSRICFIQLVSMQ-UHFFFAOYSA-N
2-trifluoromethyl quinoline, 2-trifluoromethyl-quinoline, pubchem23495, trifluoromethyl quinoline, quinoline,2-trifluoromethyl
2777805
2-(trifluoromethyl)quinoline
FC(F)(F)C1=NC2=CC=CC=C2C=C1
CAS Min % 96.0
CAS Max % 100.0
Melting Point 59.0°C to 62.0°C
Color White
Boiling Point 233.0°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 197.16
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 2-(Trifluoromethyl)quinoline
Show More Show Less
compliance-icons
Product Identifier
  • 2-(Trifluoromethyl)quinoline
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'