Alfa Aesar™ 2-(Trifluoromethyl)pyridine-5-carboxaldehyde, 95%


Overview
Brand: Alfa Aesar™

Manufacturer Part Number: H64054.MD

250MG 2-(Trifluoromethyl)pyridine-5-carboxaldehyde, 95% 250mg

UNSPSC: 12352114

Code: NEW

Additional Details:
Additional Details: CAS Number: 386704-12-7 Weight: 0.00025kg


Quantity 250mg

Product Code. 15428676

Quantity Price
1 £ 12.51 / 250mg
Estimated Shipment date
from Supplier 03-02-2017
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material 2-(Trifluoromethyl)pyridine-5-carboxaldehyde
CAS 386704-12-7
Formula Weight 175.11
MDL Number MFCD01862647
UN Number UN2811
Synonym 6-(Trifluoromethyl)nicotinaldehyde

Specification

Formula Weight 175.11
Melting Point 52° to 56°C
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material 2-(Trifluoromethyl)pyridine-5-carboxaldehyde
Physical Form Solid
Assay 95%
MDL Number MFCD01862647
CAS 386704-12-7
UN Number UN2811
Synonym 6-(Trifluoromethyl)nicotinaldehyde
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H315-H319-H317-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
Storage Note 1 2° to 8°C
DOT Information Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
Sensitivity Air Sensitive
Quantity 250mg
Safety and Handling

Danger

GHS H Statement
H301-H315-H319-H317-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

2° to 8°C