missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-(Trifluoromethyl)pyridine-5-carboxaldehyde, 95%

Brand:  Alfa Aesar™ H64054.06

Code : NEW

Additional Details : CAS Number : 386704-12-7 Weight : 0.00500kg Transport : UN number : 2811 Chem class : 6.1 Pack group : III

Quantity 5g

 View more versions of this product

Product Code. 15401089

  • £180.00 / 5g

Estimated Shipment: 26-08-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

386704-12-7
175.11
MRPAGRCGPAXOGS-UHFFFAOYSA-N
2777762
FC(F)(F)C1=NC=C(C=O)C=C1
C7H4F3NO
MFCD01862647
6-trifluoromethyl pyridine-3-carboxaldehyde, 6-trifluoromethyl nicotinaldehyde, 2-trifluoromethyl pyridine-5-carboxaldehyde, 6-trifluoromethyl pyridine-3-carbaldehyde, 5-formyl-2-trifluoromethyl pyridine, 3-pyridinecarboxaldehyde, 6-trifluoromethyl, 6-trifluoromethyl-pyridine-3-carbaldehyde, 6-trifluoromethyl-3-pyridinecarboxaldehyde, 2-trifluoromethyl pyridine-5-carbaldehyde, pubchem15866
6-(trifluoromethyl)pyridine-3-carbaldehyde

Specifications

2-(Trifluoromethyl)pyridine-5-carboxaldehyde
Air sensitive
5g
C7H4F3NO
UN2811
MRPAGRCGPAXOGS-UHFFFAOYSA-N
6-(trifluoromethyl)pyridine-3-carbaldehyde
2777762
Solid
93°C (199°F)
52°C to 56°C
386704-12-7
MFCD01862647
6-trifluoromethyl pyridine-3-carboxaldehyde, 6-trifluoromethyl nicotinaldehyde, 2-trifluoromethyl pyridine-5-carboxaldehyde, 6-trifluoromethyl pyridine-3-carbaldehyde, 5-formyl-2-trifluoromethyl pyridine, 3-pyridinecarboxaldehyde, 6-trifluoromethyl, 6-trifluoromethyl-pyridine-3-carbaldehyde, 6-trifluoromethyl-3-pyridinecarboxaldehyde, 2-trifluoromethyl pyridine-5-carbaldehyde, pubchem15866
FC(F)(F)C1=NC=C(C=O)C=C1
175.11
175.11
95%
Videos
SDS Documents
Certificates