missing translation for 'onlineSavingsMsg'
Learn More

2-(Trifluoromethyl)pyrazine, 97%, Thermo Scientific™

Catalog Number 10765462
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
61655-67-2
C5H3F3N2
148.09
MFCD06797734
CEAMSISEJZMQEP-UHFFFAOYSA-N
2-trifluoromethyl pyrazine, 2-trifluoromethyl-pyrazine, pyrazine, trifluoromethyl, trifluoromethylpyrazine, pyrazine, 2-trifluoromethyl
17888936
2-(trifluoromethyl)pyrazine
C1=CN=C(C=N1)C(F)(F)F
This item is not returnable. View return policy
61655-67-2
C5H3F3N2
148.09
MFCD06797734
CEAMSISEJZMQEP-UHFFFAOYSA-N
2-trifluoromethyl pyrazine, 2-trifluoromethyl-pyrazine, pyrazine, trifluoromethyl, trifluoromethylpyrazine, pyrazine, 2-trifluoromethyl
17888936
2-(trifluoromethyl)pyrazine
C1=CN=C(C=N1)C(F)(F)F
CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Boiling Point 116.0°C to 118.0°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 148.09
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2-(Trifluoromethyl)pyrazine
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-(Trifluoromethyl)pyrazine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'