missing translation for 'onlineSavingsMsg'
Learn More

2-(trifluoromethyl)phenyl isothiocyanate, Thermo Scientific™

Catalog Number p-4509669
Click to view available options
Quantity:
5 g
10 g
25 g
This item is not returnable. View return policy
1743-86-8
C8H4F3NS
203.182
FCEKLQPJGXIQRY-UHFFFAOYSA-N
2-trifluoromethyl phenyl isothiocyanate, 1-isothiocyanato-2-trifluoromethyl benzene, 2-trifluoromethylphenylisothiocyanate, 2-trifluoromethyl phenylisothiocyanate, 2-trifluoromethylphenyl isothiocyanate, o-trifluoromethyl phenyl isothiocyanate, 1-isothiocyanato-2-trifluoromethyl-benzene, benzene, 1-isothiocyanato-2-trifluoromethyl, alpha,alpha,alpha-trifluoro-o-tolyl isothiocyanate, isothiocyanic acid 2-trifluoromethyl phenyl ester
737162
1-isothiocyanato-2-(trifluoromethyl)benzene
C1=CC=C(C(=C1)C(F)(F)F)N=C=S
This item is not returnable. View return policy
1743-86-8
C8H4F3NS
203.182
FCEKLQPJGXIQRY-UHFFFAOYSA-N
2-trifluoromethyl phenyl isothiocyanate, 1-isothiocyanato-2-trifluoromethyl benzene, 2-trifluoromethylphenylisothiocyanate, 2-trifluoromethyl phenylisothiocyanate, 2-trifluoromethylphenyl isothiocyanate, o-trifluoromethyl phenyl isothiocyanate, 1-isothiocyanato-2-trifluoromethyl-benzene, benzene, 1-isothiocyanato-2-trifluoromethyl, alpha,alpha,alpha-trifluoro-o-tolyl isothiocyanate, isothiocyanic acid 2-trifluoromethyl phenyl ester
737162
1-isothiocyanato-2-(trifluoromethyl)benzene
C1=CC=C(C(=C1)C(F)(F)F)N=C=S
Density 1.346g/mL
Assay Percent Range 97%
Linear Formula CF3OC6H4NCS
Quantity 5 g
Formula Weight 203.19
Percent Purity 97%
Chemical Name or Material 2-(Trifluoromethyl)phenyl isothiocyanate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'