missing translation for 'onlineSavingsMsg'
Learn More

2-(Trifluoromethyl)oxazolo[4,5-b]pyridine, 97%, Thermo Scientific™

Catalog Number p-7055369
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
894406-63-4
C7H3F3N2O
188.109
MJILPMZYOJELAX-UHFFFAOYSA-N
29919200
2-(trifluoromethyl)-[1,3]oxazolo[4,5-b]pyridine
C1=CC2=C(N=C1)N=C(O2)C(F)(F)F
This item is not returnable. View return policy
894406-63-4
C7H3F3N2O
188.109
MJILPMZYOJELAX-UHFFFAOYSA-N
29919200
2-(trifluoromethyl)-[1,3]oxazolo[4,5-b]pyridine
C1=CC2=C(N=C1)N=C(O2)C(F)(F)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Synonym 2-trifluoromethyl oxazolo 4,5-b pyridine, 2-trifluoromethyl-1,3 oxazolo 4,5-b pyridine, oxazolo 4,5-b pyridine,2-trifluoromethyl, oxazolo 4,5-b pyridine, 2-trifluoromethyl, 2-trifluoromethyl 1,3 oxazolo 4,5-b pyridine
Formula Weight 188.11
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 2-(Trifluoromethyl)oxazolo[4,5-b]pyridine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'