missing translation for 'onlineSavingsMsg'
Learn More

2-(Trifluoromethyl)benzoyl chloride, 98%, Thermo Scientific Chemicals

Catalog Number p-3779761
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
312-94-7
C8H4ClF3O
208.56
MFCD00000667
MXIUWSYTQJLIKE-UHFFFAOYSA-N
2-trifluoromethyl benzoyl chloride, o-trifluoromethylbenzoyl chloride, alpha,alpha,alpha-trifluoro-o-toluoyl chloride, 2-trifluoromethyl benzoylchloride, unii-d01d0b64eq, otf-boc, 2-trifluoromethyl-benzoyl chloride, o-trifluoromethyl benzoyl chloride
67561
2-(trifluoromethyl)benzoyl chloride
FC(F)(F)C1=CC=CC=C1C(Cl)=O
This item is not returnable. View return policy
312-94-7
C8H4ClF3O
208.56
MFCD00000667
MXIUWSYTQJLIKE-UHFFFAOYSA-N
2-trifluoromethyl benzoyl chloride, o-trifluoromethylbenzoyl chloride, alpha,alpha,alpha-trifluoro-o-toluoyl chloride, 2-trifluoromethyl benzoylchloride, unii-d01d0b64eq, otf-boc, 2-trifluoromethyl-benzoyl chloride, o-trifluoromethyl benzoyl chloride
67561
2-(trifluoromethyl)benzoyl chloride
FC(F)(F)C1=CC=CC=C1C(Cl)=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point -22°C
Color Pink or Yellow
Density 1.4100g/mL
Boiling Point 84°C to 85°C (16.0 mmHg)
Flash Point 95°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CF3C6H4COCl
Refractive Index 1.4780 to 1.481
Quantity 1 g
Specific Gravity 1.41
Solubility Information Solubility in water: decomposes
Formula Weight 208.57
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2-(Trifluoromethyl)benzoyl chloride
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'