missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(Trifluoromethyl)benzenethiol, ≥95%, Thermo Scientific™
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Specifications
CAS Min % | 95.0 |
Density | 1.35g/mL |
Assay Percent Range | 95% |
Packaging | Amber Glass Bottle |
Linear Formula | CF3C6H4SH |
Quantity | 10 g |
Formula Weight | 178.18 |
Percent Purity | 95% |
Chemical Name or Material | 2-(Trifluoromethyl)benzenethiol |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'