missing translation for 'onlineSavingsMsg'
Learn More

2-(Trifluoromethyl)benzaldehyde, 97%, ACROS Organics™

Brand:  Acros Organics 232681000

Code : 29

Additional Details : CAS Number : 447-61-0 Weight : 0.15000kg


Quantity 100g
Packaging Glass bottle

 View more versions of this product

Product Code. 10577373

  • £109.20 / 100g


Estimated Shipment
09-02-2018
Add to basket


Specifications

Chemical Identifiers

2-(Trifluoromethyl)benzaldehyde
96.0
97%
C8H5F3O
174.12
α, α, α-Trifluoro-o-tolualdehyde
447-61-0
100.0
97%
CF3C6H4CHO
MFCD00003337

Specifications

2-(Trifluoromethyl)benzaldehyde
70.0° to 71.0°C (16.0 mmHg)
− 40.0°C
447-61-0
96.0
Glass bottle
Liquid
97%
CF3C6H4CHO
MFCD00003337
α, α, α-Trifluoro-o-tolualdehyde
1.3200g/mL
Clear colorless to slightly yellow
100g
Authentic
100.0
1.4650 to 1.4670
97%
C8H5F3O
174.12
07, III, 1013
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 2-(Trifluoromethyl)benzaldehyde

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/A§.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS