missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(Trifluoromethyl)aniline, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
Specifications
CAS Min % | 97.0 |
Density | 1.2960g/mL |
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Linear Formula | H2NC6H4CF3 |
Quantity | 1 g |
Formula Weight | 161.13 |
Percent Purity | 97% |
Chemical Name or Material | 2-(Trifluoromethyl)aniline |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'