missing translation for 'onlineSavingsMsg'
Learn More

2-(Trifluoromethyl)-6-pyridinecarboxylic acid, 97%, ACROS Organics™

Catalog Number 12025410
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
131747-42-7
C7H4F3NO2
191.11
MFCD02180819
OKBHXGBLXDNJJD-UHFFFAOYSA-N
6-trifluoromethyl picolinic acid, 6-trifluoromethyl pyridine-2-carboxylic acid, 2-trifluoromethyl-6-pyridinecarboxylic acid, 6-trifluoromethyl-2-pyridinecarboxylic acid, 2-pyridinecarboxylic acid, 6-trifluoromethyl, 6-trifluoromethyl pyridine-2-carboxylicacid, 6-trifluoromethyl-pyridine-2-carboxylic acid, pubchem15170, 2-trifluoromethyl-6-pyridinecarboxylicacid, acmc-1ca7g
14761453
6-(trifluoromethyl)pyridine-2-carboxylic acid
OC(=O)C1=NC(=CC=C1)C(F)(F)F
This item is not returnable. View return policy
131747-42-7
C7H4F3NO2
191.11
MFCD02180819
OKBHXGBLXDNJJD-UHFFFAOYSA-N
6-trifluoromethyl picolinic acid, 6-trifluoromethyl pyridine-2-carboxylic acid, 2-trifluoromethyl-6-pyridinecarboxylic acid, 6-trifluoromethyl-2-pyridinecarboxylic acid, 2-pyridinecarboxylic acid, 6-trifluoromethyl, 6-trifluoromethyl pyridine-2-carboxylicacid, 6-trifluoromethyl-pyridine-2-carboxylic acid, pubchem15170, 2-trifluoromethyl-6-pyridinecarboxylicacid, acmc-1ca7g
14761453
6-(trifluoromethyl)pyridine-2-carboxylic acid
OC(=O)C1=NC(=CC=C1)C(F)(F)F
Assay Percent Range 97%
Quantity 5 g
Formula Weight 191.11
Chemical Name or Material 2-(Trifluoromethyl)-6-pyridinecarboxylic acid
compliance-icons
Product Identifier
  • 2-(Trifluoromethyl)-6-pyridinecarboxylic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'