missing translation for 'onlineSavingsMsg'
Learn More

2-(Trifluoromethyl)-1H-indole, 97%, Thermo Scientific™

Catalog Number p-4380116
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
51310-54-4
C9H6F3N
185.15
QFHVHZJGQWMBTE-UHFFFAOYSA-N
2-trifluoromethyl-1h-indole, 2-trifluoromethylindole, 1h-indole, 2-trifluoromethyl, pubchem23796, 2-trifluoromethyl indole
10932124
2-(trifluoromethyl)-1H-indole
C1=CC=C2C(=C1)C=C(N2)C(F)(F)F
This item is not returnable. View return policy
51310-54-4
C9H6F3N
185.15
QFHVHZJGQWMBTE-UHFFFAOYSA-N
2-trifluoromethyl-1h-indole, 2-trifluoromethylindole, 1h-indole, 2-trifluoromethyl, pubchem23796, 2-trifluoromethyl indole
10932124
2-(trifluoromethyl)-1H-indole
C1=CC=C2C(=C1)C=C(N2)C(F)(F)F
Melting Point 107°C to 109°C
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 185.15
Percent Purity 97%
Chemical Name or Material 2-(Trifluoromethyl)-1H-indole
compliance-icons
Product Identifier
  • 2-(Trifluoromethyl)-1H-indole
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'