missing translation for 'onlineSavingsMsg'
Learn More

2-Trifluoromethoxyphenylboronic acid, 97%, Thermo Scientific Chemicals

Catalog Number 11995861
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
175676-65-0
C7H6BF3O3
205.93
MFCD01320763
AIJCNTOYZPKURP-UHFFFAOYSA-N
2-trifluoromethoxy phenylboronic acid, 2-trifluoromethoxyphenylboronic acid, 2-trifluormethoxy phenylboronic acid, 2-trifluoromethoxy benzeneboronic acid, 2-trifluoromethoxy phenyl boronic acid, 2-trifluormethoxyphenylboronic acid, o-trifluoromethoxy phenylboronic acid, 2-trifluoromethoxy phenyl boranediol, boronic acid, 2-trifluoromethoxy phenyl
2777204
[2-(trifluoromethoxy)phenyl]boronic acid
OB(O)C1=CC=CC=C1OC(F)(F)F
This item is not returnable. View return policy
175676-65-0
C7H6BF3O3
205.93
MFCD01320763
AIJCNTOYZPKURP-UHFFFAOYSA-N
2-trifluoromethoxy phenylboronic acid, 2-trifluoromethoxyphenylboronic acid, 2-trifluormethoxy phenylboronic acid, 2-trifluoromethoxy benzeneboronic acid, 2-trifluoromethoxy phenyl boronic acid, 2-trifluormethoxyphenylboronic acid, o-trifluoromethoxy phenylboronic acid, 2-trifluoromethoxy phenyl boranediol, boronic acid, 2-trifluoromethoxy phenyl
2777204
[2-(trifluoromethoxy)phenyl]boronic acid
OB(O)C1=CC=CC=C1OC(F)(F)F

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point 121.0°C to 125.0°C
Color Brown to White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass Bottle
Linear Formula F3COC6H4B(OH)2
Quantity 1 g
Formula Weight 205.93
Percent Purity 97%
Physical Form Crystalline Powder or Powder
Chemical Name or Material 2-Trifluoromethoxyphenylboronic acid
Show More Show Less
compliance-icons
Product Identifier
  • 2-(Trifluoromethoxy)phenylboronic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'