missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(Trifluoromethoxy)benzaldehyde, 96%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
Specifications
CAS Min % | 95.0 |
CAS Max % | 100.0 |
Color | Colorless to Yellow |
Boiling Point | 77°C (20.0 mmHg) |
Flash Point | 67°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 95% min. (GC) |
Quantity | 5 g |
Formula Weight | 190.12 |
Percent Purity | 96% |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'