missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(Tri-n-butylstannyl)furan, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 mL
25 mL
100 mL
Description
It is an agrochemical, organic and pharmaceutical intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is an agrochemical, organic and pharmaceutical intermediates.
Notes
Store at room temperature. Light sensitive.
It is an agrochemical, organic and pharmaceutical intermediates.
Notes
Store at room temperature. Light sensitive.
Specifications
Color | Colorless to Yellow |
Density | 1.134 g/mL |
Boiling Point | 90°C (0.1 mmHg) |
Refractive Index | 1.494 |
Quantity | 100 mL |
UN Number | UN2788 |
Formula Weight | 357.1 |
Percent Purity | 97% |
Physical Form | Liquid |
Chemical Name or Material | 2-(Tri-n-butylstannyl)furan |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'