missing translation for 'onlineSavingsMsg'
Learn More

2-Thiophenesulfonyl chloride, 98%, Thermo Scientific Chemicals

Catalog Number p-199747
37.00 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
16629-19-9
C4H3ClO2S2
182.64
MFCD00005426
VNNLHYZDXIBHKZ-UHFFFAOYSA-N
2-thiophenesulfonyl chloride, 2-thienylsulfonyl chloride, thiophene-2-sulphonyl chloride, 2-thiophenesulfonylchloride, 2-thienylsulfonylchloride, 2-thiophene sulfonyl chloride, chloro-2-thienylsulfone, pubchem7737, acmc-209dtw, thiophenesulfonyl chloride
85518
thiophene-2-sulfonyl chloride
ClS(=O)(=O)C1=CC=CS1
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
16629-19-9
C4H3ClO2S2
182.64
MFCD00005426
VNNLHYZDXIBHKZ-UHFFFAOYSA-N
2-thiophenesulfonyl chloride, 2-thienylsulfonyl chloride, thiophene-2-sulphonyl chloride, 2-thiophenesulfonylchloride, 2-thienylsulfonylchloride, 2-thiophene sulfonyl chloride, chloro-2-thienylsulfone, pubchem7737, acmc-209dtw, thiophenesulfonyl chloride
85518
thiophene-2-sulfonyl chloride
ClS(=O)(=O)C1=CC=CS1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 27°C to 29°C
Boiling Point 80°C (1.0 mmHg)
Flash Point >113°C
Assay Percent Range 97.5% min. (Ex Cl) (Argentometry)
Packaging Glass bottle
Refractive Index 1.5722 to 1.5742
Quantity 5 g
Formula Weight 182.65
Percent Purity 98%
Chemical Name or Material 2-Thiophenesulfonyl chloride, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 2-Thiophenesulfonyl chloride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'