missing translation for 'onlineSavingsMsg'
Learn More

2-Thiophenecarboxylic acid, 99%, Thermo Scientific Chemicals

Catalog Number p-3758816
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
527-72-0
C5H4O2S
128.15
MFCD00005437
QERYCTSHXKAMIS-UHFFFAOYSA-N
2-thiophenecarboxylic acid, 2-thenoic acid, 2-carboxythiophene, 2-thiophenic acid, 2-thienylcarboxylic acid, alpha-thiophenecarboxylic acid, 2-tca, thiophene-2-carboxylate, tenoic acid, .alpha.-thiophenecarboxylic acid
10700
CHEBI:71241
thiophene-2-carboxylic acid
OC(=O)C1=CC=CS1
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
527-72-0
C5H4O2S
128.15
MFCD00005437
QERYCTSHXKAMIS-UHFFFAOYSA-N
2-thiophenecarboxylic acid, 2-thenoic acid, 2-carboxythiophene, 2-thiophenic acid, 2-thienylcarboxylic acid, alpha-thiophenecarboxylic acid, 2-tca, thiophene-2-carboxylate, tenoic acid, .alpha.-thiophenecarboxylic acid
10700
CHEBI:71241
thiophene-2-carboxylic acid
OC(=O)C1=CC=CS1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point 126.0°C to 130.0°C
Boiling Point 260.0°C
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Quantity 5 g
Beilstein 18,289
Merck Index 15,9507
Solubility Information Solubility in water: 80g/L (20°C). Other solubilities: soluble in alcohol and ether, moderately soluble in chcl3, slightly soluble in petroleum ether
Formula Weight 128.15
Percent Purity 99%
Chemical Name or Material 2-Thiophenecarboxylic acid
Show More Show Less
compliance-icons
Product Identifier
  • 2-Thiophenecarboxylic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'