missing translation for 'onlineSavingsMsg'
Learn More

2-Thiophenecarboxamide, 99%, Thermo Scientific™

Catalog Number p-3758834
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
5813-89-8
C5H5NOS
127.17
MFCD00010425
DENPQNAWGQXKCU-UHFFFAOYSA-N
2-thiophenecarboxamide, 2-thienylamide, 2-thenamide, thiophen-2-carboxamide, 2-aminocarbonyl thiophene, 2-thienylamide french, thiophenecarboxamide, thiolamide, acmc-1ao1m, 5-18-06-00162 beilstein handbook reference
22063
thiophene-2-carboxamide
C1=CSC(=C1)C(=O)N
This item is not returnable. View return policy
5813-89-8
C5H5NOS
127.17
MFCD00010425
DENPQNAWGQXKCU-UHFFFAOYSA-N
2-thiophenecarboxamide, 2-thienylamide, 2-thenamide, thiophen-2-carboxamide, 2-aminocarbonyl thiophene, 2-thienylamide french, thiophenecarboxamide, thiolamide, acmc-1ao1m, 5-18-06-00162 beilstein handbook reference
22063
thiophene-2-carboxamide
C1=CSC(=C1)C(=O)N
CAS Min % 98.5
CAS Max % 100.0
Melting Point 179.0°C to 182.0°C
Color Gray to White
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 127.17
Percent Purity 99%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 2-Thiophenecarboxamide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'