missing translation for 'onlineSavingsMsg'
Learn More

2-Thiophenecarboxaldehyde 98%, ACROS Organics™

Brand:  Acros Organics 138831000

Code : 29

Additional Details : CAS Number : 98-03-3 Weight : 0.17000kg


Quantity 100mL
Packaging Glass bottle

 View more versions of this product

Product Code. 10684992

  • £29.60 / 100mL


EU Stock 12
Add to basket


Specifications

Chemical Identifiers

2-Thiophenecarboxaldehyde
98-03-3
100.0
98%
112.15
98%
97.5
98%
C5H4OS
MFCD00005429

Specifications

2-Thiophenecarboxaldehyde
98-03-3
100.0
Liquid
98%
Glass bottle
MFCD00005429
Solubility in water: insoluble. Other solubilities: soluble in alcohol, ether and benzene
Product may darken on standing
Clear yellow to light brown
100mL
98%
97.5
Authentic
98%
C5H4OS
112.15
1.5890 to 1.5920
1.2
1.2000g/mL
198.0°C
77°C
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 2-Thiophenecarboxaldehyde

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2

Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS