missing translation for 'onlineSavingsMsg'
Learn More

2-Thiopheneacetic acid, 98%, Thermo Scientific Chemicals

Catalog Number p-199646
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass Bottle
Plastic bottle
This item is not returnable. View return policy
1918-77-0
MFCD00005458
SMJRBWINMFUUDS-UHFFFAOYSA-N
2-thiopheneacetic acid, 2-thienylacetic acid, thiopheneacetic acid, 2-thiophen-2-yl acetic acid, thiophene-2-acetic acid, 2-2-thienyl acetic acid, 2-thiophene acetic acid, thiophen-2-ylacetic acid, thien-2-yl acetic acid, 2-carboxymethyl thiophene
15970
CHEBI:45807
2-thiophen-2-ylacetic acid
C1=CSC(=C1)CC(=O)O
This item is not returnable. View return policy
1918-77-0
MFCD00005458
SMJRBWINMFUUDS-UHFFFAOYSA-N
2-thiopheneacetic acid, 2-thienylacetic acid, thiopheneacetic acid, 2-thiophen-2-yl acetic acid, thiophene-2-acetic acid, 2-2-thienyl acetic acid, 2-thiophene acetic acid, thiophen-2-ylacetic acid, thien-2-yl acetic acid, 2-carboxymethyl thiophene
15970
CHEBI:45807
2-thiophen-2-ylacetic acid
C1=CSC(=C1)CC(=O)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 61°C to 65°C
Color Brown
Boiling Point 160°C (22.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass Bottle
Molecular Formula C6H6O2S
Quantity 5 g
Beilstein 18, 293
Molecular Weight (g/mol) 142.17
Formula Weight 142.17
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 2-Thiopheneacetic acid, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 2-Thiopheneacetic acid
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'