missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Thiopheneacetic acid, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
500 g
Description
2-Thiopheneacetic acid is used in the preparation of rosette-like nanoscale gold materials. Further, it is used as a pharmaceutical intermediate for cefaloridine and cefalotin sodium.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Thiopheneacetic acid is used in the preparation of rosette-like nanoscale gold materials. Further, it is used as a pharmaceutical intermediate for cefaloridine and cefalotin sodium.
Solubility
Soluble in water, ethanol, ether and carbon tetrachloride.
Notes
Incompatible with strong oxidizing agents and strong bases.
2-Thiopheneacetic acid is used in the preparation of rosette-like nanoscale gold materials. Further, it is used as a pharmaceutical intermediate for cefaloridine and cefalotin sodium.
Solubility
Soluble in water, ethanol, ether and carbon tetrachloride.
Notes
Incompatible with strong oxidizing agents and strong bases.
Specifications
Melting Point | 61°C to 65°C |
Boiling Point | 160°C (22 mmHg) |
Quantity | 25 g |
UN Number | UN3261 |
Beilstein | 114551 |
Solubility Information | Soluble in water,ethanol,ether and carbon tetrachloride. |
Formula Weight | 142.18 |
Percent Purity | 98% |
Chemical Name or Material | 2-Thiopheneacetic acid |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'