missing translation for 'onlineSavingsMsg'
Learn More

2-Thionaphthol, 98+%, Thermo Scientific Chemicals

Catalog Number p-7081923
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy
91-60-1
C10H8S
160.234
MFCD00004086
RFCQDOVPMUSZMN-UHFFFAOYSA-N
2-naphthalenethiol, 2-mercaptonaphthalene, 2-thionaphthol, thionaphthol, 2-naphthyl mercaptan, vulcamel tbn, 2-naphthyl thiol, renacit 1, rpa no. 2, rpa 2
7058
naphthalene-2-thiol
C1=CC=C2C=C(C=CC2=C1)S
This item is not returnable. View return policy
91-60-1
C10H8S
160.234
MFCD00004086
RFCQDOVPMUSZMN-UHFFFAOYSA-N
2-naphthalenethiol, 2-mercaptonaphthalene, 2-thionaphthol, thionaphthol, 2-naphthyl mercaptan, vulcamel tbn, 2-naphthyl thiol, renacit 1, rpa no. 2, rpa 2
7058
naphthalene-2-thiol
C1=CC=C2C=C(C=CC2=C1)S

2-Thionaphthol was used in the preparation of cholesterol monolayer and multilayer Langmuir- Blodgett (LB) films. The magnetic resonance shift for a self-assembled monolayer of 2-naphthalenethiol was studied that suggested considerable promise in flexible and transparent photonic devices for biological and chemical sensing.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Thionaphthol was used in the preparation of cholesterol monolayer and multilayer Langmuir- Blodgett (LB) films. The magnetic resonance shift for a self-assembled monolayer of 2-naphthalenethiol was studied that suggested considerable promise in flexible and transparent photonic devices for biological and chemical sensing.

Solubility
Soluble in alcohol, dipropylene glycol. Soluble in water 34.4 mg/L @ 25°C.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Air sensitive. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 79°C to 83°C
Density 1.55
Boiling Point 286°C
Quantity 10 g
Beilstein 636389
Sensitivity Air Sensitive
Merck Index 14,6379
Solubility Information Soluble in alcohol,dipropylene glycol. Soluble in water 34.4mg/L @ 25°C.
Formula Weight 160.24
Percent Purity ≥98%
Chemical Name or Material 2-Thionaphthol
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'