missing translation for 'onlineSavingsMsg'
Learn More

2-Thiomorpholinoisonicotinonitrile, 97%, Thermo Scientific™

Catalog Number p-3522464
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
884507-30-6
C10H11N3S
205.28
MFCD09025862
MZIANIKUVGCEPB-UHFFFAOYSA-N
2-thiomorpholinoisonicotinonitrile, 2-thiomorpholin-4-yl pyridine-4-carbonitrile, 4-pyridinecarbonitrile,2-4-thiomorpholinyl, 2-1,4-thiazaperhydroin-4-yl pyridine-4-carbonitrile
18525850
2-(thiomorpholin-4-yl)pyridine-4-carbonitrile
N#CC1=CC(=NC=C1)N1CCSCC1
This item is not returnable. View return policy
884507-30-6
C10H11N3S
205.28
MFCD09025862
MZIANIKUVGCEPB-UHFFFAOYSA-N
2-thiomorpholinoisonicotinonitrile, 2-thiomorpholin-4-yl pyridine-4-carbonitrile, 4-pyridinecarbonitrile,2-4-thiomorpholinyl, 2-1,4-thiazaperhydroin-4-yl pyridine-4-carbonitrile
18525850
2-(thiomorpholin-4-yl)pyridine-4-carbonitrile
N#CC1=CC(=NC=C1)N1CCSCC1
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 205.28
Percent Purity 97%
Chemical Name or Material 2-Thiomorpholinoisonicotinonitrile
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'