Learn More
Thermo Scientific Maybridge 2-Thienylboronic acid, 100g, 97%
Brand: Thermo Scientific Maybridge CC13012FA
2-Thienylboronic acid, 100g, 97%
Code : 31
Additional Details : CAS Number : 6165-68-0 Weight : 0.15000kg
Brand: Thermo Scientific Maybridge CC13012FA
2-Thienylboronic acid, 100g, 97%
Code : 31
Additional Details : CAS Number : 6165-68-0 Weight : 0.15000kg