missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Maybridge 2-Thienyl isocyanate, 10g, 90%

Brand:  Thermo Scientific Maybridge CC13006EA

2-Thienyl isocyanate, 10g, 90%

Code : 31

Additional Details : CAS Number : 2048-57-9 Weight : 0.06000kg Transport : UN number : 2206 Chem class : 6.1 Pack group : III

Product Code. 10673393

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.