Learn More
Thermo Scientific Maybridge 2-Thienyl isocyanate, 5g, 90%
Brand: Thermo Scientific Maybridge CC13006DE
2-Thienyl isocyanate, 5g, 90%
Code : 31
Additional Details : CAS Number : 2048-57-9 Weight : 0.05500kg Transport : UN number : 2206 Chem class : 6.1 Pack group : III