missing translation for 'onlineSavingsMsg'
Learn More

(2-Thien-2-ylphenyl)methanol, 97%, Thermo Scientific™

Catalog Number p-3109870
Click to view available options
Quantity:
250 mg
1 g
10 g
This item is not returnable. View return policy
773872-97-2
C11H10OS
190.26
MFCD06203081
IUULMJAEIKVTKZ-UHFFFAOYSA-N
2-thien-2-ylphenyl methanol, 2-thiophen-2-yl phenyl methanol, 2-thien-2-yl phenyl methanol, 2-thiophen-2-ylphenyl methanol, 2-2-thienyl phenyl methanol, benzenemethanol,2-2-thienyl, 2-2-thienyl phenyl methan-1-ol
2795559
OCC1=CC=CC=C1C1=CC=CS1
This item is not returnable. View return policy
773872-97-2
C11H10OS
190.26
MFCD06203081
IUULMJAEIKVTKZ-UHFFFAOYSA-N
2-thien-2-ylphenyl methanol, 2-thiophen-2-yl phenyl methanol, 2-thien-2-yl phenyl methanol, 2-thiophen-2-ylphenyl methanol, 2-2-thienyl phenyl methanol, benzenemethanol,2-2-thienyl, 2-2-thienyl phenyl methan-1-ol
2795559
OCC1=CC=CC=C1C1=CC=CS1
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 250 mg
IUPAC Name (2-thiophen-2-ylphenyl)methanol
Formula Weight 190.27
Percent Purity 97%
Chemical Name or Material (2-Thien-2-ylphenyl)methanol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'