missing translation for 'onlineSavingsMsg'
Learn More

2-Thenylmercaptan, 98%, Thermo Scientific Chemicals

Catalog Number p-3779670
92.00 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
6258-63-5
C5H6S2
130.22
MFCD00107135
GCZQHDFWKVMZOE-UHFFFAOYSA-N
2-thiophenemethanethiol, thenyl mercaptan, 2-thenylmercaptan, 2-thienylmethanethiol, thiophene-2-methanethiol, 2-thenylthiol, thiophen-2-yl-methanethiol, unii-9jml279xs6, 2-thenyl mercaptan, 2-mercaptomethyl thiophene
80408
thiophen-2-ylmethanethiol
C1=CSC(=C1)CS
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
6258-63-5
C5H6S2
130.22
MFCD00107135
GCZQHDFWKVMZOE-UHFFFAOYSA-N
2-thiophenemethanethiol, thenyl mercaptan, 2-thenylmercaptan, 2-thienylmethanethiol, thiophene-2-methanethiol, 2-thenylthiol, thiophen-2-yl-methanethiol, unii-9jml279xs6, 2-thenyl mercaptan, 2-mercaptomethyl thiophene
80408
thiophen-2-ylmethanethiol
C1=CSC(=C1)CS

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Density 1.2000g/mL
Boiling Point 90°C (15.0 mmHg)
Flash Point 81°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Refractive Index 1.6020 to 1.604
Quantity 1 g
Specific Gravity 1.2
Solubility Information Solubility in water: slightly soluble
Formula Weight 130.22
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2-Thenylmercaptan
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'