missing translation for 'onlineSavingsMsg'
Learn More

[2-(Tetrahydropyran-4-yloxy)phenyl]methylamine, 97%, Maybridge

Brand:  Maybridge CC52213DA

Code : 31

Additional Details : CAS Number : 898289-33-3 Weight : 0.05100kg Transport : UN number : 2735 Chem class : 8 Pack group : III

Quantity 1g
Packaging Amber Glass Bottle

 View more versions of this product

Product Code. 10275614

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Specifications

Chemical Identifiers

[2-(Tetrahydropyran-4-yloxy)phenyl]methylamine
96.5
97%
C12H17NO2
MFCD09025871
898289-33-3
100.0
97%
207.27

Specifications

[2-(Tetrahydropyran-4-yloxy)phenyl]methylamine
898289-33-3
100.0
97%
C12H17NO2
MFCD09025871
1g
96.5
Amber Glass Bottle
97%
207.27
Videos
SDS