missing translation for 'onlineSavingsMsg'
Learn More

2-tert-Butyl-4-methylphenol 99%, ACROS Organics™

Brand:  Acros Organics 107965000

Code : 29

Additional Details : CAS Number : 2409-55-4 Weight : 0.55000kg Transport : UN number : 2430 Chem class : 1a Pack group : III


Quantity 500g
Packaging Glass bottle

 View more versions of this product

Product Code. 10569990

  • £68.50 / 500g


EU Stock 4
Add to basket


Specifications

Chemical Identifiers

2-tert-Butyl-4-methylphenol
2409-55-4
100.0
99%
(CH3)3CC6H3(CH3)OH
MFCD00002381
99%
98.5
99%
C11H16O
164.25
2-tert-Butyl-p-cresol

Specifications

2-tert-Butyl-4-methylphenol
2409-55-4
100.0
Crystalline Mass
99%
(CH3)3CC6H3(CH3)OH
164.25
2-tert-Butyl-p-cresol
237.0°C
White to yellow-beige
105°C
99%
98.5
Authentic
99%
C11H16O
Glass bottle
MFCD00002381
Solubility in water: insoluble. Other solubilities: soluble in aceton, benzene, chloroform
50.0° to 53.0°C
500g
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 2-tert-Butyl-4-methylphenol

Signal Word
  • Danger

Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H318-Causes serious eye damage.
  • H411-Toxic to aquatic life with long lasting effects.
  • H314-Causes severe skin burns and eye damage.

Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/A§.

SDS