missing translation for 'onlineSavingsMsg'
Learn More

2-TEDC, Tocris Bioscience™

5-, 12-, 15-Lipoxygenase inhibitor

Brand:  Tocris (Bio-Techne) 064550

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13467843

  • £554.19 / 50mg

Estimated Shipment date
from Supplier 06-04-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Potent inhibitor of 5-, 12-, and 15-lipoxygenase (IC50 values are 0.09, 0.013 and 0.5 μM respectively).
Specifications

Chemical Identifiers

132465-10-2
315.343
2-tedc, chembl37081, tocris-0645, 2-thiophen-2-yl ethyl 2-cyano-3-3,4-dihydroxyphenyl prop-2-enoate, e-2-cyano-3-3,4-dihydroxyphenyl acrylic acid 2-2-thienyl ethyl ester, 2-cyano-3-3,4-dihydroxy-phenyl-acrylic acid 2-thiophen-2-yl-ethyl ester, 2-cyano-3-3,4-dihydroxyphenyl propenoic acid 2-2-thienyl ethyl ester, 2-thiophen-2-ylethyl e-2-cyano-3-3,4-dihydroxyphenyl prop-2-enoate, 2-propenoic acid,2-cyano-3-3,4-dihydroxyphenyl-,2-2-thienyl ethyl ester, e-9ci
2-thiophen-2-ylethyl (E)-2-cyano-3-(3,4-dihydroxyphenyl)prop-2-enoate
C16H13NO4S
KCGLTUBCAGZLHP-XYOKQWHBSA-N
6069552
C1=CSC(=C1)CCOC(=O)C(=CC2=CC(=C(C=C2)O)O)C#N

Specifications

50mg
132465-10-2
2-tedc, chembl37081, tocris-0645, 2-thiophen-2-yl ethyl 2-cyano-3-3,4-dihydroxyphenyl prop-2-enoate, e-2-cyano-3-3,4-dihydroxyphenyl acrylic acid 2-2-thienyl ethyl ester, 2-cyano-3-3,4-dihydroxy-phenyl-acrylic acid 2-thiophen-2-yl-ethyl ester, 2-cyano-3-3,4-dihydroxyphenyl propenoic acid 2-2-thienyl ethyl ester, 2-thiophen-2-ylethyl e-2-cyano-3-3,4-dihydroxyphenyl prop-2-enoate, 2-propenoic acid,2-cyano-3-3,4-dihydroxyphenyl-,2-2-thienyl ethyl ester, e-9ci
C1=CSC(=C1)CCOC(=O)C(=CC2=CC(=C(C=C2)O)O)C#N
315.343
315.34
2-TEDC
C16H13NO4S
KCGLTUBCAGZLHP-XYOKQWHBSA-N
2-thiophen-2-ylethyl (E)-2-cyano-3-(3,4-dihydroxyphenyl)prop-2-enoate
6069552
>99%
Videos
Documents