missing translation for 'onlineSavingsMsg'
Learn More

2-(Styrylthio)acetic acid, 97%, Thermo Scientific™

Catalog Number 10535934
Click to view available options
Quantity:
1 g
10 g
25 g
This item is not returnable. View return policy
13435-97-7
C10H10O2S
194.248
MFCD00068078
IUEVBKDVPSQVLM-VOTSOKGWSA-N
2-styrylthio acetic acid, e-2-phenylethenyl sulfanyl acetic acid, 2-phenylethenyl sulfanyl acetic acid, acetic acid,2-2-phenylethenyl thio, e-styrylsulfanyl-acetic acid, 2-1e-2-phenylvinylthio acetic acid, 2-e-2-phenylethenyl sulfanylacetic acid
5474136
2-[(E)-2-phenylethenyl]sulfanylacetic acid
C1=CC=C(C=C1)C=CSCC(=O)O
This item is not returnable. View return policy
13435-97-7
C10H10O2S
194.248
MFCD00068078
IUEVBKDVPSQVLM-VOTSOKGWSA-N
2-styrylthio acetic acid, e-2-phenylethenyl sulfanyl acetic acid, 2-phenylethenyl sulfanyl acetic acid, acetic acid,2-2-phenylethenyl thio, e-styrylsulfanyl-acetic acid, 2-1e-2-phenylvinylthio acetic acid, 2-e-2-phenylethenyl sulfanylacetic acid
5474136
2-[(E)-2-phenylethenyl]sulfanylacetic acid
C1=CC=C(C=C1)C=CSCC(=O)O
CAS Min % 95.0
Assay Percent Range 95%
Linear Formula C6H5CH=CHSCH2CO2H
Quantity 25 g
Formula Weight 194.25
Percent Purity 95%
Chemical Name or Material 2-(Styrylthio)acetic acid
compliance-icons
Product Identifier
  • 2-(Styrylthio)acetic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'