Learn More
Thermo Scientific Maybridge 2-Quinoxalinecarboxylic acid, 1g, 97%
Brand: Thermo Scientific Maybridge CC03501DA
2-Quinoxalinecarboxylic acid, 1g, 97%
Code : 31
Additional Details : CAS Number : 879-65-2 Weight : 0.05100kg
Brand: Thermo Scientific Maybridge CC03501DA
2-Quinoxalinecarboxylic acid, 1g, 97%
Code : 31
Additional Details : CAS Number : 879-65-2 Weight : 0.05100kg