missing translation for 'onlineSavingsMsg'
Learn More

2-Quinoxalinecarboxylic acid, 97%, Thermo Scientific™

Catalog Number p-2690146
Click to view available options
Quantity:
100 mg
500 mg
Packaging:
Glass bottle
This item is not returnable. View return policy
879-65-2
C9H6N2O2
174.16
MFCD00012334
UPUZGXILYFKSGE-UHFFFAOYSA-N
2-quinoxalinecarboxylic acid, 2-quinoxalinecarboxylicacid, unii-m5oe8sn42m, 2-carboxyquinoxaline, m5oe8sn42m, 2-qca, pubchem9776, carbadox metabolite, acmc-1bl7g, quinoxaline carboxylic acid
96695
quinoxaline-2-carboxylic acid
OC(=O)C1=CN=C2C=CC=CC2=N1
This item is not returnable. View return policy
879-65-2
C9H6N2O2
174.16
MFCD00012334
UPUZGXILYFKSGE-UHFFFAOYSA-N
2-quinoxalinecarboxylic acid, 2-quinoxalinecarboxylicacid, unii-m5oe8sn42m, 2-carboxyquinoxaline, m5oe8sn42m, 2-qca, pubchem9776, carbadox metabolite, acmc-1bl7g, quinoxaline carboxylic acid
96695
quinoxaline-2-carboxylic acid
OC(=O)C1=CN=C2C=CC=CC2=N1
CAS Min % 96.0
CAS Max % 100.0
Melting Point 206.0°C
Color Beige to Green-Yellow
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 100 mg
Formula Weight 174.16
Percent Purity 97%
Physical Form Powder, Crystals or Chunks
Chemical Name or Material 2-Quinoxalinecarboxylic acid
Show More Show Less
compliance-icons
Product Identifier
  • 2-Quinoxalinecarboxylic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'