missing translation for 'onlineSavingsMsg'
Learn More

2-Quinoxalinecarbaldehyde, 97+%, Maybridge

Brand:  Maybridge CC03504EA

Code : 31

Additional Details : CAS Number : 1593-08-4 Weight : 0.06000kg

Quantity 10g
Packaging Amber Glass Bottle

 View more versions of this product

Product Code. 10547464

  • £421.00 / 10g


Estimated Shipment date
from Supplier 26-02-2018
Add to basket

Specifications

Chemical Identifiers

2-Quinoxalinecarbaldehyde
97%
158.16
97%
C9H6N2O

Specifications

2-Quinoxalinecarbaldehyde
Amber Glass Bottle
97%
158.16
1593-08-4
97%
C9H6N2O
10g
Videos
SDS