missing translation for 'onlineSavingsMsg'
Learn More

2-Quinolinecarbonitrile, 97%, Thermo Scientific Chemicals

Catalog Number 13428678
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
1436-43-7
C10H6N2
154.17
MFCD00134341
WDXARTMCIRVMAE-UHFFFAOYSA-N
2-quinolinecarbonitrile, quinaldonitrile, 2-cyanoquinoline, quinolinecarbonitrile, cyanoquinoline, 2-cyano-quinoline, quinoline-2-carbinitrile, acmc-1bzd0
74031
quinoline-2-carbonitrile
N#CC1=CC=C2C=CC=CC2=N1
This item is not returnable. View return policy
1436-43-7
C10H6N2
154.17
MFCD00134341
WDXARTMCIRVMAE-UHFFFAOYSA-N
2-quinolinecarbonitrile, quinaldonitrile, 2-cyanoquinoline, quinolinecarbonitrile, cyanoquinoline, 2-cyano-quinoline, quinoline-2-carbinitrile, acmc-1bzd0
74031
quinoline-2-carbonitrile
N#CC1=CC=C2C=CC=CC2=N1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 93°C to 95°C
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 154.17
Percent Purity 97%
Chemical Name or Material 2-Quinolinecarbonitrile
compliance-icons
Product Identifier
  • 2-Quinolinecarbonitrile
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'