missing translation for 'onlineSavingsMsg'
Learn More

2-Pyrrolidinone, 99%, Thermo Scientific Chemicals

Catalog Number 10489440
Click to view available options
Quantity:
250 mL
500 mL
2.5 L
Packaging:
Glass bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
616-45-5
C4H7NO
85.11
MFCD00005270
HNJBEVLQSNELDL-UHFFFAOYSA-N
2-pyrrolidinone, pyrrolidone, butyrolactam, 2-pyrrolidone, 2-oxopyrrolidine, pyrrolidon, 2-ketopyrrolidine, 2-pyrol, pyrrolidinone, gamma-butyrolactam
12025
CHEBI:36592
pyrrolidin-2-one
C1CC(=O)NC1
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
616-45-5
C4H7NO
85.11
MFCD00005270
HNJBEVLQSNELDL-UHFFFAOYSA-N
2-pyrrolidinone, pyrrolidone, butyrolactam, 2-pyrrolidone, 2-oxopyrrolidine, pyrrolidon, 2-ketopyrrolidine, 2-pyrol, pyrrolidinone, gamma-butyrolactam
12025
CHEBI:36592
pyrrolidin-2-one
C1CC(=O)NC1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point 25.5°C
Color Colorless to Yellow
Density 1.1030g/mL
Boiling Point 250.0°C
Flash Point 138°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4860 to 1.4880
Quantity 500 mL
Beilstein 21, 236
Merck Index 15, 8130
Specific Gravity 1.103
Solubility Information Solubility in water: miscible. Other solubilities: miscible with ethanol, ether, chloroform, benzene,, ethyl acetate, carbon disulfide
Viscosity 10.2 mPa.s (30°C)
Formula Weight 85.11
Percent Purity 99%
Physical Form Liquid or Low Melting Mass
Chemical Name or Material 2-Pyrrolidinone, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 2-Pyrrolidinone
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'