(2-Pyrrolidin-1-ylphenyl)methylamine 97+%, Maybridge


Overview
Brand: Maybridge

Manufacturer Part Number: CC44813DA

UNSPSC: 12350000

Code: 31

Additional Details:
Additional Details: CAS Number: 72752-53-5 Weight: 0.05100kg Transport: UN number : 2735 Chem class : 1b Pack group : III


Packaging Amber Glass Bottle
Quantity 1g

Product Code. 10271643

Quantity Price
1 £ 72.0 / 1g
Estimated Shipment date
from Supplier 07-06-2017
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material (2-Pyrrolidin-1-ylphenyl)methylamine
Assay 95%
Assay Percent Range 95%
Molecular Formula C11H16N2
Formula Weight 176.26

Specification

Formula Weight 176.26
Quantity 1g
Chemical Name or Material (2-Pyrrolidin-1-ylphenyl)methylamine
Assay 95%
Molecular Formula C11H16N2
Assay Percent Range 95%
Packaging Amber Glass Bottle
CAS 72752-53-5
Documentation

Technical documents