missing translation for 'onlineSavingsMsg'
Learn More

2-Pyridinethioamide, 97%, Thermo Scientific Chemicals

Catalog Number p-4228893
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
5346-38-3
C6H6N2S
138.19
MFCD00087576
HYKQYVSNFPWGKQ-UHFFFAOYSA-N
thiopicolinamide, 2-pyridinecarbothioamide, pyridine-2-thioamide, pyridine-2-carbothioic acid amide, picolinamide, thio, 2-pyridinethiocarboxamide, picolinothioamide, 2-thiopicolinamide, pyridinecarbothioamide, 2-thiocarbamoylpyridine
1549499
pyridine-2-carbothioamide
NC(=S)C1=CC=CC=N1
This item is not returnable. View return policy
5346-38-3
C6H6N2S
138.19
MFCD00087576
HYKQYVSNFPWGKQ-UHFFFAOYSA-N
thiopicolinamide, 2-pyridinecarbothioamide, pyridine-2-thioamide, pyridine-2-carbothioic acid amide, picolinamide, thio, 2-pyridinethiocarboxamide, picolinothioamide, 2-thiopicolinamide, pyridinecarbothioamide, 2-thiocarbamoylpyridine
1549499
pyridine-2-carbothioamide
NC(=S)C1=CC=CC=N1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point 137°C to 139°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 138.19
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 2-Pyridinethioamide
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-Pyridinethioamide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H315-Causes skin irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'