missing translation for 'onlineSavingsMsg'
Learn More

2-Pyridin-3-ylaniline, 95%, Maybridge

Brand:  Maybridge CC41814DA

Code : 31

Additional Details : CAS Number : 177202-83-4 Weight : 0.05100kg Transport : UN number : 2811 Chem class : 6.1 Pack group : III

Packaging Amber Glass Bottle
Quantity 1g

 View more versions of this product

Product Code. 10507104

  • £131.00 / 1g

Estimated Shipment date
from Supplier 19-05-2020

Please to purchase this item. Need a web account? Register with us today!


Specifications

Chemical Identifiers

177202-83-4
170.215
2-pyridin-3-yl aniline, 2-3-pyridyl aniline, 2-pyridin-3-yl-phenylamine, 2-pyridin-3-ylphenylamine, benzenamine, 2-3-pyridinyl, 2-3-pyridyl phenylamine, 3-o-aminophenyl pyridine, 3-2-aminophenyl pyridine
2-pyridin-3-ylaniline
C11H10N2
MTYHJHATMPCNLW-UHFFFAOYSA-N
459519
C1=CC=C(C(=C1)C2=CN=CC=C2)N

Specifications

2-Pyridin-3-ylaniline
1g
95%
2-pyridin-3-yl aniline, 2-3-pyridyl aniline, 2-pyridin-3-yl-phenylamine, 2-pyridin-3-ylphenylamine, benzenamine, 2-3-pyridinyl, 2-3-pyridyl phenylamine, 3-o-aminophenyl pyridine, 3-2-aminophenyl pyridine
C1=CC=C(C(=C1)C2=CN=CC=C2)N
170.215
170.21
Amber Glass Bottle
177202-83-4
C11H10N2
MTYHJHATMPCNLW-UHFFFAOYSA-N
2-pyridin-3-ylaniline
459519
95%
Videos
SDS Documents