missing translation for 'onlineSavingsMsg'
Learn More

2-Pyrazinecarboxylic acid, 99%, Thermo Scientific Chemicals

Catalog Number p-188787
Click to view available options
Quantity:
25 g
100 g
Packaging:
Plastic bottle
This item is not returnable. View return policy
98-97-5
C5H4N2O2
124.1
MFCD00006130
NIPZZXUFJPQHNH-UHFFFAOYSA-N
2-pyrazinecarboxylic acid, pyrazinoic acid, pyrazinecarboxylic acid, pyrazinic acid, 2-carboxypyrazine, paradiazinecarboxylic acid, pyrazinemonocarboxylic acid, 2-pyrazinoic acid, 2-pyrazine carboxylic acid, piazinecarboxylic acid
1047
CHEBI:71311
pyrazine-2-carboxylic acid
C1=CN=C(C=N1)C(=O)O
This item is not returnable. View return policy
98-97-5
C5H4N2O2
124.1
MFCD00006130
NIPZZXUFJPQHNH-UHFFFAOYSA-N
2-pyrazinecarboxylic acid, pyrazinoic acid, pyrazinecarboxylic acid, pyrazinic acid, 2-carboxypyrazine, paradiazinecarboxylic acid, pyrazinemonocarboxylic acid, 2-pyrazinoic acid, 2-pyrazine carboxylic acid, piazinecarboxylic acid
1047
CHEBI:71311
pyrazine-2-carboxylic acid
C1=CN=C(C=N1)C(=O)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point 222.0°C to 225.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Plastic bottle
Quantity 25 g
Beilstein 25, 125
Merck Index 15, 8070
Solubility Information Solubility in water: soluble in cold water. Other solubilities: 1g/120g absolubte ethanol (25 c),practically insoluble in ether,chloroform and,benzene
Formula Weight 124.1
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 2-Pyrazinecarboxylic acid, 99%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'