missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Pyrazinecarbonyl chloride, Technical Grade, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Specifications
Packaging | Amber Glass Bottle |
Quantity | 5 g |
Formula Weight | 142.54 |
Percent Purity | Tech. |
Grade | Technical |
Chemical Name or Material | 2-Pyrazinecarbonyl chloride |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'