missing translation for 'onlineSavingsMsg'
Learn More

2-Propanol, Spectrophotometric Grade, 99.7+%, Thermo Scientific Chemicals

Catalog Number p-7050646
Click to view available options
Quantity:
1 L
4 x 1 L
This item is not returnable. View return policy
67-63-0
C3H8O
60.096
MFCD00011674
KFZMGEQAYNKOFK-UHFFFAOYSA-N
isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol
3776
CHEBI:17824
propan-2-ol
CC(C)O
This item is not returnable. View return policy
67-63-0
C3H8O
60.096
MFCD00011674
KFZMGEQAYNKOFK-UHFFFAOYSA-N
isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol
3776
CHEBI:17824
propan-2-ol
CC(C)O

Solvent for gums, resins, alkaloids, essential oils. Denaturant for ethyl alcohol. In antifreeze compositions. It is used in lotions and cosmetics. Isopropyl alcohol is used in keyboard, LCD and laptop cleaning. It is used as a wetting agent in the fountain solution used in lithographic printing, and often used as a solvent for French polishing shellac used in cabinet making. It act as hydride source in the Meerwein-Ponndorf-Verley reduction

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -90°C
Density 0.785 g/mL
Boiling Point 82°C to 83°C
Quantity 1 L
Flash Point 12°C (53°F)
Odor Alcohol-like
Physical Form Liquid
Refractive Index 1.377
UN Number UN1219
Beilstein 635639
Merck Index 14,5208
Solubility Information Miscible with water,ethanol,acetone,chloroform,methanol,diethyl ether,n-octanol and benzene.
Formula Weight 60.1
Percent Purity ≥99.7%
Grade Spectrophotometric
Chemical Name or Material 2-Propanol
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'