missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Propanol, Puriss. p.a., ACS Reagent, ≥99.8% (GC), Honeywell Riedel-de Haën™
Click to view available options
Quantity:
1 L
2.5 L
25 L
Unit Size:
1 L
2.5 L
25 L
Specifications
Specifications
| Density | 0.785g/mL at 20°C, 0.785g/mL at 25°C |
| Quantity | 25 L |
| Explosion Limit | 0.02 |
| Flash Point | 12°C (53.6°F) |
| Ignition Point | 750°F |
| Linear Formula | (CH3)2CHOH |
| Vapor Pressure | 44 mmHg (25°C), 33 mmHg (20°C) |
| UN Number | UN1219 |
| Beilstein | 635639 |
| Formula Weight | 60.1g/mol |
| Show More |
Safety and Handling
Product Identifier
- 2-Propanol
- Danger
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
- Specific target organ toxicity Category 3
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction