missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Isopropanol, Optima™LC/MS Grade, Fisher Chemical™
29.79 GBP valid until 2024-03-30
Use promo code "21663" to get your promotional price.
31.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.
Use promo code "21663" to get your promotional price.
31.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.
Click to view available options
Quantity:
500 mL
1 L
2.5 L
4 L
Packaging:
Glass Bottle
Wide Neck Amber Bottle
Description
2-Propanol, >-99.90%, C3H8O, CAS Number-67-63-0, petrohol, isopropyl alcohol, isopropanol, sec-propyl alcohol, dimethylcarbinol, 2-hydroxypropane, hartosol, 2-propanol, alkolave, avantine, 4L, Report AU at 254nm, Report AU at 210nm, Report AU at 230nm, Report AU at 220nm
Specifications
pH | 7 |
Melting Point | -89.5°C |
Absorbance | Report AU at 210nm, Report AU at 220nm, Report AU at 230nm, Report AU at 254nm |
Color | Colorless |
Density | 0.785g/cm³ |
Boiling Point | 81°C |
Quantity | 500 mL |
Packaging | Glass Bottle |
Physical Form | Liquid |
Assay Percent Range | Assay: ≥99.90% |
Show More |
Safety and Handling
Product Identifier
- Iso-propanol
- Danger
- Flammable liquid Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P240-Ground/bond container and receiving equipment.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'