missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Propanol, BAKER ANALYZED PHOTREX™, J.T. Baker™
Click to view available options
Quantity:
1 L
Packaging:
Glass
Description
PHOTREX™ solvents are manufactured to minimize lot-to-lot variability and contaminants that can interfere with UV, and in some cases IR, spectra, including residue after evaporation, and acid and base concentrations. Function testing confirms maximum absorbance in selected wavelengths; 50-100% transmittance windows in IR wavelengths are reported.
- High quality chemicals for laboratory and specialized industrial use
- Exceeds the requirements set forth by the American Chemical Society (ACS)
- Label provides an actual Lot Analysis as a guarantee of purity and consistency
- Analytical methods used for these determinations are based on standard test procedures described in technical publications
- Improvements developed by J.T. Baker are employed to achieve a higher degree of reliability
Specifications
Melting Point | -89.5°C |
Absorbance | Max. 1.00 at 210nm, Max. 0.40 at 220nm, Max. 0.20 at 230nm, Max. 0.08 at 245nm, Max. 0.04 (at 260nm) - 0.01 (at 330-400nm) |
CAS | 67-63-0 |
Molecular Formula | C3H8O |
Linear Formula | CH3CHOHCH3 |
Synonym | isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol |
InChI Key | KFZMGEQAYNKOFK-UHFFFAOYSA-N |
SMILES | CC(C)O |
IUPAC Name | propan-2-ol |
Molecular Weight (g/mol) | 60.096 |
Show More |
Safety and Handling
Signal Word
- Danger
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 3
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H226-Flammable liquid and vapour.
- H318-Causes serious eye damage.
- H315-Causes skin irritation.
- H335-May cause respiratory irritation.
- H336-May cause drowsiness or dizziness.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'